Assay Details
| Gene Symbol: eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002427.2 | NP_001002427.1 | 2 - 3 | 194 | 88 | |
| GenBank mRNA | BC076140.1 | - | 2 - 3 | 143 | 88 | |
More Information
Set Membership: |
|

eny2
Genomic Map