Assay Details
| Gene Symbol: etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034175.1 | NP_001029347.1 | 1 - 2 | 341 | 57 | |
| GenBank mRNA | BC096922.1 | - | 1 - 2 | 341 | 57 | |
| BC164400.1 | - | 1 - 2 | 182 | 57 | ||
More Information
Set Membership: |
|

etnk2
Genomic Map