Assay Details
| Gene Symbol: shox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126411.1 | NP_001119883.1 | 1 - 2 | 366 | 91 | |
| XM_005167854.4 | XP_005167911.1 | 1 - 2 | 392 | 91 | ||
| GenBank mRNA | BC107618.1 | - | 1 - 2 | 377 | 91 | |
More Information
Set Membership: |
|

shox
Genomic Map