Assay Details
Gene Symbol: grid2ipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044903.1 | NP_001038368.1 | 11 - 12 | 2449 | 124 | |
XM_009306633.3 | XP_009304908.1 | 18 - 19 | 3847 | 124 | ||
XM_017358463.2 | XP_017213952.2 | 18 - 19 | 3835 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
