Assay Details
Gene Symbol: grid2ipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044903.1 | NP_001038368.1 | 1 - 2 | 287 | 83 | |
XM_009306633.3 | XP_009304908.1 | 5 - 6 | 1286 | 83 | ||
XM_017358463.2 | XP_017213952.2 | 5 - 6 | 1286 | 83 | ||
XM_021480122.1 | XP_021335797.1 | 5 - 6 | 1417 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
