Assay Details
| Gene Symbol: grid2ipb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044903.1 | NP_001038368.1 | 6 - 7 | 1195 | 64 | |
| XM_009306633.3 | XP_009304908.1 | 13 - 14 | 2548 | 64 | ||
| XM_017358463.2 | XP_017213952.2 | 13 - 14 | 2548 | 64 | ||
| XM_021480122.1 | XP_021335797.1 | 13 - 14 | 2679 | 64 | ||
More Information
Set Membership: |
|

grid2ipb
Genomic Map