Assay Details
| Gene Symbol: tmem106bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045052.1 | NP_001038517.1 | 6 - 7 | 673 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem106bb| Gene Symbol: tmem106bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045052.1 | NP_001038517.1 | 6 - 7 | 673 | 76 | |
Set Membership: |
|