Assay Details
| Gene Symbol: ccnd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045068.1 | NP_001038533.1 | 1 - 2 | 190 | 137 | |
| XM_005164521.4 | XP_005164578.1 | 1 - 2 | 461 | 137 | ||
| GenBank mRNA | FJ648074.1 | - | 1 - 2 | 253 | 137 | |
More Information
Set Membership: |
|

ccnd2b
Genomic Map