Assay Details
| Gene Symbol: rab9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242974.1 | NP_001229903.1 | 4 - 5 | 417 | 66 | |
| GenBank mRNA | BC122264.1 | - | 4 - 5 | 401 | 66 | |
| BC165268.1 | - | 1 - 2 | 185 | 66 | ||
| CB360735.1 | - | 4 - 5 | 340 | 66 | ||
More Information
Set Membership: |
|

rab9a
Genomic Map