Assay Details
| Gene Symbol: lratd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044880.2 | NP_001038345.1 | 1 | 1264 | 93 | |
| GenBank mRNA | BC122257.1 | - | 1 | 845 | 93 | |
More Information
Set Membership: |
|

lratd2a
Genomic Map