Assay Details
| Gene Symbol: zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077291.2 | NP_001070759.2 | 3 - 4 | 431 | 78 | |
| XM_005160177.4 | XP_005160234.1 | 3 - 4 | 522 | 78 | ||
| XM_009294848.3 | XP_009293123.1 | 3 - 4 | 522 | 78 | ||
| XM_009294859.3 | XP_009293134.1 | 3 - 4 | 522 | 78 | ||
| GenBank mRNA | BC124649.1 | - | 3 - 4 | 431 | 78 | |
More Information
Set Membership: |
|

zfyve27
Genomic Map