Assay Details
| Gene Symbol: fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077303.1 | NP_001070771.1 | 2 - 3 | 184 | 115 | |
| GenBank mRNA | BC124831.1 | - | 2 - 3 | 184 | 115 | |
More Information
Set Membership: |
|

fblim1
Genomic Map