Assay Details
| Gene Symbol: deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077320.1 | NP_001070788.2 | 7 - 8 | 1257 | 65 | |
| XM_005173523.4 | XP_005173580.1 | 7 - 8 | 1319 | 65 | ||
| XM_005173524.4 | XP_005173581.1 | 7 - 8 | 1336 | 65 | ||
| GenBank mRNA | BC124609.1 | - | 7 - 8 | 1257 | 65 | |
More Information
Set Membership: |
|

deptor
Genomic Map