Assay Details
| Gene Symbol: dele1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080086.1 | NP_001073555.1 | 896 | 100 | ||
| GenBank mRNA | BC128827.1 | - | 896 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dele1| Gene Symbol: dele1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080086.1 | NP_001073555.1 | 896 | 100 | ||
| GenBank mRNA | BC128827.1 | - | 896 | 100 | ||
Set Membership: |
|