Assay Details
| Gene Symbol: dele1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080086.1 | NP_001073555.1 | 6 - 7 | 1054 | 115 | |
| GenBank mRNA | BC128827.1 | - | 6 - 7 | 1054 | 115 | |
More Information
Set Membership: |
|

dele1
Genomic Map