Assay Details
| Gene Symbol: snapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080163.2 | NP_001073632.1 | 2 - 3 | 170 | 138 | |
| XM_005159085.4 | XP_005159142.1 | 2 - 3 | 468 | 138 | ||
| GenBank mRNA | BC133151.1 | - | 2 - 3 | 170 | 138 | |
More Information
Set Membership: |
|

snapc5
Genomic Map