Assay Details
| Gene Symbol: bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080653.1 | NP_001074122.1 | 4 - 5 | 492 | 66 | |
| GenBank mRNA | BC129445.1 | - | 4 - 5 | 492 | 66 | |
More Information
Set Membership: |
|

bckdhb
Genomic Map