Assay Details
| Gene Symbol: ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080663.2 | NP_001074132.2 | 2 - 3 | 696 | 74 | |
| GenBank mRNA | BC129188.1 | - | 2 - 3 | 701 | 74 | |
More Information
Set Membership: |
|

ribc1
Genomic Map