Assay Details
| Gene Symbol: trpm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080678.2 | NP_001074147.2 | 7 - 8 | 1074 | 85 | |
| NM_001316373.1 | NP_001303302.1 | 6 - 7 | 957 | 85 | ||
| XM_017356917.2 | XP_017212406.1 | 7 - 8 | 1256 | 85 | ||
| XM_017356918.2 | XP_017212407.1 | 7 - 8 | 1256 | 85 | ||
| XM_017356919.2 | XP_017212408.1 | 7 - 8 | 1103 | 85 | ||
| GenBank mRNA | BC129336.1 | - | 7 - 8 | 1074 | 85 | |
| BC164017.1 | - | 5 - 6 | 778 | 85 | ||
| KF305306.1 | - | 6 - 7 | 953 | 85 | ||
More Information
Set Membership: |
|

trpm1a
Genomic Map