Assay Details
| Gene Symbol: ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082921.1 | NP_001076390.1 | 2 - 3 | 952 | 71 | |
| GenBank mRNA | BC133878.1 | - | 2 - 3 | 952 | 71 | |
More Information
Set Membership: |
|

ppp1r15a
Genomic Map