Assay Details
| Gene Symbol: tnfrsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089510.2 | NP_001082979.2 | 4 - 5 | 541 | 134 | |
| XM_005155914.4 | XP_005155971.1 | 4 - 5 | 536 | 134 | ||
| GenBank mRNA | BC134212.1 | - | 4 - 5 | 541 | 134 | |
| HM239645.1 | - | 3 - 4 | 320 | 134 | ||
More Information
Set Membership: |
|

tnfrsf1b
Genomic Map