Assay Details
| Gene Symbol: ube2j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089535.1 | NP_001083004.1 | 2 - 3 | 289 | 60 | |
| GenBank mRNA | BC139531.1 | - | 2 - 3 | 289 | 60 | |
More Information
Set Membership: |
|

ube2j2
Genomic Map