Assay Details
| Gene Symbol: lrrc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110372.1 | NP_001103842.1 | 3 - 4 | 1184 | 62 | |
| GenBank mRNA | BC093410.1 | - | 3 - 4 | 824 | 62 | |
| BC133856.1 | - | 3 - 4 | 1103 | 62 | ||
| BC154501.1 | - | 3 - 4 | 1184 | 62 | ||
More Information
Set Membership: |
|

lrrc47
Genomic Map