Assay Details
| Gene Symbol: mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111238.1 | NP_001104708.1 | 3 - 4 | 634 | 62 | |
| GenBank mRNA | BC155626.1 | - | 3 - 4 | 634 | 62 | |
| BC171376.1 | - | 3 - 4 | 598 | 62 | ||
More Information
Set Membership: |
|

mri1
Genomic Map