Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 17 - 18 | 2192 | 78 | |
| XM_005157694.4 | XP_005157751.1 | 9 - 10 | 955 | 78 | ||
| XM_009291655.3 | XP_009289930.1 | 16 - 17 | 3249 | 78 | ||
| XM_017350981.2 | XP_017206470.1 | 16 - 17 | 3247 | 78 | ||
| GenBank mRNA | AM749802.1 | - | 17 - 18 | 2192 | 78 | |
More Information
Set Membership: |
|

nos2b
Genomic Map