Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 19 - 20 | 2462 | 69 | |
| XM_005157694.4 | XP_005157751.1 | 11 - 12 | 1225 | 69 | ||
| XM_009291655.3 | XP_009289930.1 | 18 - 19 | 3519 | 69 | ||
| XM_017350981.2 | XP_017206470.1 | 18 - 19 | 3517 | 69 | ||
| GenBank mRNA | AM749802.1 | - | 19 - 20 | 2462 | 69 | |
More Information
Set Membership: |
|

nos2b
Genomic Map