Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 1 - 2 | 181 | 74 | |
| GenBank mRNA | AM749802.1 | - | 1 - 2 | 181 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AM749802.1, NM_001113501.1 transcripts.
Set Membership: |
|

nos2b
Genomic Map