Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 23 - 24 | 3045 | 67 | |
| XM_005157694.4 | XP_005157751.1 | 15 - 16 | 1808 | 67 | ||
| XM_009291655.3 | XP_009289930.1 | 22 - 23 | 4080 | 67 | ||
| XM_017350981.2 | XP_017206470.1 | 22 - 23 | 4100 | 67 | ||
| GenBank mRNA | AM749802.1 | - | 23 - 24 | 3045 | 67 | |
| EU332350.1 | - | 23 - 24 | 2896 | 67 | ||
More Information
Set Membership: |
|

nos2b
Genomic Map