Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 24 - 25 | 3190 | 112 | |
| XM_005157694.4 | XP_005157751.1 | 16 - 17 | 1953 | 112 | ||
| XM_017350981.2 | XP_017206470.1 | 23 - 24 | 4245 | 112 | ||
| GenBank mRNA | AM749802.1 | - | 24 - 25 | 3190 | 112 | |
| EU332350.1 | - | 24 - 25 | 3041 | 112 | ||
More Information
Set Membership: |
|

nos2b
Genomic Map