Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 4 - 5 | 505 | 84 | |
| XM_009291655.3 | XP_009289930.1 | 3 - 4 | 1562 | 84 | ||
| XM_017350981.2 | XP_017206470.1 | 3 - 4 | 1560 | 84 | ||
| GenBank mRNA | AM749802.1 | - | 4 - 5 | 505 | 84 | |
| EU332350.1 | - | 4 - 5 | 356 | 84 | ||
More Information
Set Membership: |
|

nos2b
Genomic Map