Assay Details
| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 6 - 7 | 762 | 109 | |
| XM_009291655.3 | XP_009289930.1 | 5 - 6 | 1819 | 109 | ||
| XM_017350981.2 | XP_017206470.1 | 5 - 6 | 1817 | 109 | ||
| GenBank mRNA | AM749802.1 | - | 6 - 7 | 762 | 109 | |
| EU332350.1 | - | 6 - 7 | 613 | 109 | ||
More Information
Set Membership: |
|

nos2b
Genomic Map