Assay Details
| Gene Symbol: cadm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113551.2 | NP_001107023.2 | 9 - 10 | 1069 | 118 | |
| XM_009295329.3 | XP_009293604.1 | 8 - 9 | 1106 | 118 | ||
| GenBank mRNA | EU182351.2 | - | 9 - 10 | 1069 | 118 | |
More Information
Set Membership: |
|

cadm1a
Genomic Map