Assay Details
| Gene Symbol: cadm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113552.1 | NP_001107024.1 | 5 - 6 | 599 | 100 | |
| GenBank mRNA | CU638788.1 | - | 5 - 6 | 543 | 100 | |
| EU182352.1 | - | 5 - 6 | 599 | 100 | ||
More Information
Set Membership: |
|

cadm1b
Genomic Map