Assay Details
| Gene Symbol: anxa5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113582.1 | NP_001107054.1 | 8 - 9 | 585 | 72 | |
| GenBank mRNA | BC154565.1 | - | 8 - 9 | 585 | 72 | |
More Information
Set Membership: |
|

anxa5a
Genomic Map