Assay Details
| Gene Symbol: setmar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113600.2 | NP_001107072.1 | 1 - 2 | 190 | 98 | |
| GenBank mRNA | BC154590.1 | - | 1 - 2 | 191 | 98 | |
| BC171571.1 | - | 1 - 2 | 190 | 98 | ||
| BC171599.1 | - | 1 - 2 | 190 | 98 | ||
More Information
Set Membership: |
|

setmar
Genomic Map