Assay Details
| Gene Symbol: crlf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113602.1 | NP_001107074.1 | 1 - 2 | 81 | 87 | |
| GenBank mRNA | BC154596.1 | - | 1 - 2 | 81 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC154596.1, NM_001113602.1 transcripts.
Set Membership: |
|

crlf1b
Genomic Map