Assay Details
| Gene Symbol: crlf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113602.1 | NP_001107074.1 | 8 - 9 | 1317 | 124 | |
| XM_017353164.2 | XP_017208653.1 | 7 - 8 | 1791 | 124 | ||
| GenBank mRNA | BC154596.1 | - | 8 - 9 | 1317 | 124 | |
More Information
Set Membership: |
|

crlf1b
Genomic Map