Assay Details
| Gene Symbol: ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113650.1 | NP_001107122.1 | 2 - 3 | 295 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ndufaf2| Gene Symbol: ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113650.1 | NP_001107122.1 | 2 - 3 | 295 | 67 | |
Set Membership: |
|