Assay Details
| Gene Symbol: zgc:165600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113661.1 | 249 | 103 | |||
| GenBank mRNA | BC154416.1 | - | 249 | 103 | ||
| BC171722.1 | - | 232 | 103 | |||
More Information
Set Membership: |
|

zgc:165600
Genomic Map