Assay Details
| Gene Symbol: fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114348.1 | NP_001107820.1 | 2 - 3 | 1216 | 66 | |
| GenBank mRNA | AB207804.1 | - | 2 - 3 | 1216 | 66 | |
| BC124439.1 | - | 2 - 3 | 1214 | 66 | ||
More Information
Set Membership: |
|

fezf1
Genomic Map