Assay Details
| Gene Symbol: exd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114406.1 | NP_001107878.1 | 8 - 9 | 1362 | 66 | |
| XM_005169892.3 | XP_005169949.1 | 8 - 9 | 1414 | 66 | ||
| GenBank mRNA | BC068411.1 | - | 7 - 8 | 928 | 66 | |
| BC159253.1 | - | 8 - 9 | 1362 | 66 | ||
More Information
Set Membership: |
|

exd2
Genomic Map