Assay Details
| Gene Symbol: mfsd4ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114416.1 | NP_001107888.1 | 4 - 5 | 873 | 74 | |
| GenBank mRNA | BC155330.1 | - | 4 - 5 | 873 | 74 | |
More Information
Set Membership: |
|

mfsd4ab
Genomic Map