Assay Details
| Gene Symbol: mfsd4ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114416.1 | NP_001107888.1 | 8 - 9 | 1390 | 96 | |
| GenBank mRNA | BC090783.1 | - | 1 - 2 | 49 | 96 | |
| BC155330.1 | - | 8 - 9 | 1390 | 96 | ||
More Information
Set Membership: |
|

mfsd4ab
Genomic Map