Assay Details
| Gene Symbol: cip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114421.1 | NP_001107893.1 | 10 - 11 | 1599 | 63 | |
| XM_005162708.4 | XP_005162765.1 | 10 - 11 | 1648 | 63 | ||
| XM_021470191.1 | XP_021325866.1 | 11 - 12 | 1338 | 63 | ||
| GenBank mRNA | BC155117.1 | - | 10 - 11 | 1599 | 63 | |
More Information
Set Membership: |
|

cip2a
Genomic Map