Assay Details
| Gene Symbol: cdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131109.2 | NP_571184.2 | 1 - 2 | 643 | 75 | |
| GenBank mRNA | BC076488.1 | - | 1 - 2 | 618 | 75 | |
| BC154241.1 | - | 1 - 2 | 637 | 75 | ||
| BC160643.1 | - | 1 - 2 | 643 | 75 | ||
| BC164427.1 | - | 1 - 2 | 435 | 75 | ||
| X66958.1 | - | 583 | 75 | |||
More Information
Set Membership: |
|

cdx4
Genomic Map