Assay Details
| Gene Symbol: dnajc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131630.1 | NP_571705.1 | 2 - 3 | 330 | 90 | |
| GenBank mRNA | AF295376.1 | - | 2 - 3 | 330 | 90 | |
More Information
Set Membership: |
|

dnajc3b
Genomic Map