Assay Details
| Gene Symbol: dnajc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131630.1 | NP_571705.1 | 10 - 11 | 1208 | 71 | |
| GenBank mRNA | AF295376.1 | - | 10 - 11 | 1208 | 71 | |
| BC163418.1 | - | 9 - 10 | 1119 | 71 | ||
| BC163421.1 | - | 9 - 10 | 1119 | 71 | ||
More Information
Set Membership: |
|

dnajc3b
Genomic Map