Assay Details
| Gene Symbol: cxadr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152948.1 | NP_694480.1 | 2 - 3 | 334 | 58 | |
| GenBank mRNA | AF268197.1 | - | 2 - 3 | 334 | 58 | |
| BC045286.1 | - | 2 - 3 | 391 | 58 | ||
| BC164203.1 | - | 2 - 3 | 257 | 58 | ||
| BC171370.1 | - | 2 - 3 | 269 | 58 | ||
| BC171372.1 | - | 2 - 3 | 269 | 58 | ||
| CU458753.1 | - | 2 - 3 | 264 | 58 | ||
More Information
Set Membership: |
|

cxadr
Genomic Map