Assay Details
| Gene Symbol: cxadr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152948.1 | NP_694480.1 | 6 - 7 | 954 | 77 | |
| GenBank mRNA | AF268197.1 | - | 6 - 7 | 954 | 77 | |
| BC045286.1 | - | 6 - 7 | 1011 | 77 | ||
| BC164203.1 | - | 6 - 7 | 877 | 77 | ||
| BC171370.1 | - | 6 - 7 | 889 | 77 | ||
| BC171372.1 | - | 6 - 7 | 889 | 77 | ||
| CU458753.1 | - | 6 - 7 | 884 | 77 | ||
More Information
Set Membership: |
|

cxadr
Genomic Map