Assay Details
| Gene Symbol: scinla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178131.3 | NP_835232.2 | 15 - 16 | 2020 | 72 | |
| GenBank mRNA | BC125898.1 | - | 15 - 16 | 1991 | 72 | |
| BC134865.1 | - | 15 - 16 | 2020 | 72 | ||
More Information
Set Membership: |
|

scinla
Genomic Map